Learn More
Biomatik Corporation Human Galanin-Like Peptide (Galp) Elisa Kit
Supplier:  Biomatik Corporation EKC33775
Encompass_Preferred
Human Galanin-Like Peptide (Galp) Elisa Kit for detecting Galanin-Like Peptide in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UBC7; GALP. Target Synonyms: GALP; Galanin-like peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3984