Learn More
Biomatik Corporation Human Glucose-6-Phosphate Isomerase, Gpi Elisa Kit
Supplier:  Biomatik Corporation EKC33833
Encompass_Preferred
Human Glucose-6-Phosphate Isomerase, Gpi Elisa Kit for detecting Glucose Phosphate Isomerase in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800ng/ml. Sensitivity: 3.12ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06744; GPI. Target Synonyms: AMF; Aurocrine motility factor; Autocrine motility factor; DKFZp686C13233; EC 5.3.1.9; G6PI_HUMAN; Glucose phosphate isomerase; Glucose-6-phosphate isomerase; GNPI; GPI; Gpi1; Hexose monophosphate isomerase; Hexosephosphate isomerase; Neuroleukin; NLK; Oxoisomerase; PGI; PHI; Phosphoglucose isomerase; Phosphohexomutase; Phosphohexose isomerase; Phosphosaccharomutase; SA 36; SA-36; SA36; Sperm antigen 36. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3992