Learn More
Biomatik Corporation Human Glutathione S-Transferase Omega-1 (Gsto1) Elisa Kit
Supplier:  Biomatik Corporation EKC33860
Encompass_Preferred
Human Glutathione S-Transferase Omega-1 (Gsto1) Elisa Kit for detecting Glutathione S-Transferase Omega 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P78417; GSTO1. Target Synonyms: AA407097; AI194287; AU018802; DKFZp686H13163; EC 2.5.1.18; Glutathione S transferase omega 1; Glutathione S-transferase omega 1-1; Glutathione S-transferase omega; Glutathione S-transferase omega-1; Glutathione transferase omega 1; Glutathione-dependent dehydroascorbate reductase; Glutathione-S-transferase like; Glutathione-S-transferase omega 1; GSTO 1-1; GSTO-1; GSTO1 1; Gsto1; GSTO1_HUMAN; GSTTLp28; GSTX; Gtsttl; MGC94845; MMA (V) reductase; Monomethylarsonic acid reductase; OTTHUMP00000020429; p28; S-(Phenacyl)glutathione reductase; SPG-R. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4000