Learn More
Biomatik Corporation Human Glyceraldehyde-3-Phosphate Dehydrogenase (Gapdh/G3Pdh) Elisa Kit
Supplier:  Biomatik Corporation EKC33866
Encompass_Preferred
Human Glyceraldehyde-3-Phosphate Dehydrogenase (Gapdh/G3Pdh) Elisa Kit for detecting Glyceraldehyde-3-Phosphate Dehydrogenase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04406; GAPDH. Target Synonyms: 38 kDa BFA-dependent ADP-ribosylation substrate; aging associated gene 9 protein; Aging-associated gene 9 protein; BARS-38; cb609; EC 1.2.1.12; Epididymis secretory sperm binding protein Li 162eP; G3P_HUMAN; G3PD; G3PDH; GAPD; GAPDH; Glyceraldehyde 3 phosphate dehydrogenase; glyceraldehyde 3-PDH; Glyceraldehyde-3-phosphate dehydrogenase; HEL-S-162eP; KNC-NDS6; MGC102544; MGC102546; MGC103190; MGC103191; MGC127711; MGC88685; OCAS; p38 component; OCT1 coactivator in S phase; 38-KD component; peptidyl cysteine S nitrosylase GAPDH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4002