Learn More
Biomatik Corporation Human Glypican 6 (Gpc6) Elisa Kit
Supplier:  Biomatik Corporation EKC33881
Encompass_Preferred
Human Glypican 6 (Gpc6) Elisa Kit for detecting Glypican 6 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y625; GPC6. Target Synonyms: A Kit; Glypican 6 [Precursor]; Glypican proteoglycan 6; Gpc6; GPC6_HUMAN; MGC126288; OMIMD1; PRO705; Secreted glypican 6; Secreted glypican-6; UNQ369. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4006