Learn More
Biomatik Corporation Human Golgi Protein 73, Gp-73 Elisa Kit
Supplier:  Biomatik Corporation EKC33887
Encompass_Preferred
Human Golgi Protein 73, Gp-73 Elisa Kit for detecting Golgi Membrane Protein 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10IU/ml. Sensitivity: 0.039IU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8NBJ4; GOLM1. Target Synonyms: bA379P1.3; C9orf155; Chromosome 9 open reading frame 155; Golgi membrane protein 1; Golgi membrane protein GP73; Golgi phosphoprotein 2; Golgi protein 73 kD; Golgi protein 73kD; GOLM 1; GOLM1; GOLM1_HUMAN; GOLPH 2; GOLPH2; GP 73; GP73; PSEC0257. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4008