Learn More
Biomatik Corporation Human Group Xiia Secretory Phospholipase A2 (Pla2G12A) Elisa Kit
Supplier:  Biomatik Corporation EKC33909
Encompass_Preferred
Human Group Xiia Secretory Phospholipase A2 (Pla2G12A) Elisa Kit for detecting Phospholipase A2, Group Xiia in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 4.7-300pg/ml. Sensitivity: 1.2pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q9BZM1; PLA2G12A. Target Synonyms: EC 3.1.1.4; Group XII secreted phospholipase A2; Group XIIA secreted phospholipase A2; Group XIIA secretory phospholipase A2; Group XIIA secretory phospholipase A2 precursor; GXII; GXII sPLA2; PG12A_HUMAN; phosphatidylcholine 2 acylhydrolase 12A; Phosphatidylcholine 2 acylhydrolase GXII; Phosphatidylcholine 2-acylhydrolase 12A; Phospholipase A2 group XIIA; PLA2G12; Pla2g12a; ROSSY; sPLA2 XII; sPLA2-XII. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4014