Learn More
Biomatik Corporation Human Heart Fatty Acid Binding Protein, H-Fabp Elisa Kit
Supplier:  Biomatik Corporation EKC33942
Encompass_Preferred
Human Heart Fatty Acid Binding Protein, H-Fabp Elisa Kit for detecting Fatty Acid Binding Protein 3, Muscle And Heart (Mammary-Derived Growth Inhibitor) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05413; FABP3. Target Synonyms: 422 protein; Cardiac FABP; Cardiac Fatty Acid Binding Protein; FABP 11; FABP 3; FABP11; FABP3; FABPH_HUMAN; Fatty acid binding protein 11; Fatty acid binding protein 3; Fatty acid binding protein 3 muscle and heart; Fatty acid binding protein 3 muscle and heart mammary derived growth inhibitor; Fatty acid binding protein 3 muscle; Fatty acid binding protein 3; muscle and heart (mammary derived growth inhibitor); Fatty acid binding protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4019