Learn More
Biomatik Corporation Human Heat Shock Protein Beta-2 (Hspb2) Elisa Kit
Supplier:  Biomatik Corporation EKC33956
Encompass_Preferred
Human Heat Shock Protein Beta-2 (Hspb2) Elisa Kit for detecting Heat Shock 27Kda Protein 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q16082; HSPB2. Target Synonyms: DMPK-binding protein; Heat shock 27kD protein 2; Heat shock 27kDa protein 2; Heat shock protein beta-2; Heat-shock protein beta-2; Hs.78846; HSP27; HspB2; HSPB2_HUMAN; LOH11CR1K; MGC133245; MKBP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4023