Learn More
Biomatik Corporation Human Histone Rna Hairpin-Binding Protein (Slbp) Elisa Kit
Supplier:  Biomatik Corporation EKC34051
Encompass_Preferred
Human Histone Rna Hairpin-Binding Protein (Slbp) Elisa Kit for detecting Stem-Loop Binding Protein in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q14493; SLBP. Target Synonyms: Hairpin binding protein histone; HBP; Histone binding protein; Histone RNA hairpin binding protein; Histone RNA hairpin-binding protein; Histone stem-loop-binding protein; SLBP; SLBP_HUMAN; Stem loop (histone) binding protein; Stem loop binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4035