Learn More
Biomatik Corporation Human Hydroxyacid Oxidase 1 (Hao1) Elisa Kit
Supplier:  Biomatik Corporation EKC34089
Encompass_Preferred
Human Hydroxyacid Oxidase 1 (Hao1) Elisa Kit for detecting Hydroxyacid Oxidase (Glycolate Oxidase) 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.69pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UJM8; HAO1. Target Synonyms: (S) 2 hydroxy acid oxidase; Glycolate oxidase; GOX; GOX1; HAO1; HAOX1; HAOX1_HUMAN; Hydroxyacid oxidase 1; MGC142225; MGC142227; OTTHUMP00000030231. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4041