Learn More
Biomatik Corporation Human Ig gamma-3 chain C region(IGHG3) ELISA kit
Supplier:  Biomatik Corporation EKC34103
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 2.7 ng/mL-2000 ng/mL. Sensitivity: 2.7 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01860. Target Synonyms: DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, MGC45809,. Restriction: For Research Use Only.
Catalog No. 50-229-4045