Learn More
Biomatik Corporation Human Inhibitor Of Growth Protein 4 (Ing4) Elisa Kit
Supplier:  Biomatik Corporation EKC34132
Encompass_Preferred
Human Inhibitor Of Growth Protein 4 (Ing4) Elisa Kit for detecting Inhibitor Of Growth Family, Member 4 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 47-3000pg/ml. Sensitivity: 11.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UNL4; ING4. Target Synonyms: Brain my036 protein; Candidate tumor suppressor p33 ING 1 homolog; Candidate tumor suppressor p33 ING1 homolog; D6Wsu147e; D6Xrf92; ING 1 like protein; ING 4; ING1 like protein; ING4; ING4_HUMAN; Inhibitor of growth family member 4; Inhibitor of growth family member 4 long isoform; Inhibitor of growth protein 4; MGC12557; my036; p29 ING 4; p29 ING4; p29ING4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4050