Learn More
Biomatik Corporation Human Integrin Beta-7 (Itgb7) Elisa Kit
Supplier:  Biomatik Corporation EKC34174
Encompass_Preferred
Human Integrin Beta-7 (Itgb7) Elisa Kit for detecting Integrin, Beta 7 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P26010; ITGB7. Target Synonyms: ITGB7; Integrin beta-7; Gut homing receptor beta subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4057