Learn More
Biomatik Corporation Human Interferon Alpha-Inducible Protein 27, Mitochondrial (Ifi27) Elisa Kit
Supplier:  Biomatik Corporation EKC34195
Encompass_Preferred
Human Interferon Alpha-Inducible Protein 27, Mitochondrial (Ifi27) Elisa Kit for detecting Interferon, Alpha-Inducible Protein 27 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 28-1800pg/ml. Sensitivity: 7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P40305; IFI27. Target Synonyms: IFI27; Interferon alpha-inducible protein 27; mitochondrial; p27; Interferon alpha-induced 11.5 kDa protein; Interferon-stimulated gene 12a protein; ISG12 (a; ISG12A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4062