Learn More
Biomatik Corporation Human Interferon Kappa (Ifnk) Elisa Kit
Supplier:  Biomatik Corporation EKC34200
Encompass_Preferred
Human Interferon Kappa (Ifnk) Elisa Kit for detecting Interferon, Kappa in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9P0W0; IFNK. Target Synonyms: IFN kappa; IFN-kappa; Ifnk; IFNK_HUMAN; Interferon kappa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4064