Learn More
Biomatik Corporation Human Interleukin 31(IL-31) ELISA Kit
Supplier:  Biomatik Corporation EKC34245
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 125 pg/mL-8000 pg/mL. Sensitivity: 31.25 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q6EBC2. Target Synonyms: IL-31. Restriction: For Research Use Only.
Catalog No. 50-229-4068