Learn More
Biomatik Corporation Human Interleukin-31 Receptor Subunit Alpha (Il31Ra) Elisa Kit
Supplier:  Biomatik Corporation EKC34297
Encompass_Preferred
Human Interleukin-31 Receptor Subunit Alpha (Il31Ra) Elisa Kit for detecting Interleukin 31 Receptor A in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8NI17; IL31RA. Target Synonyms: Class I cytokine receptor; CRL 3; CRL; CRL3; Cytokine receptor like 3; Cytokine receptor-like 3; GLM R; GLM-R; GLMR; Gp130 like monocyte receptor; GP130 like receptor; Gp130-like monocyte receptor; Gp130-like receptor; GPL; HGLM R; HGLM-R; HGLMR; IL 31 receptor subunit alpha; IL 31R subunit alpha; IL 31RA; IL-31 receptor subunit alpha; IL-31R subunit alpha; IL-31R-alpha; IL-31RA; IL31 receptor subunit alpha; IL31R subunit alpha; IL31R_HUMAN; IL31RA; Interleukin 31 receptor A; interleukin 31 receptor subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4075