Learn More
Biomatik Corporation Human Kelch-Like Ech-Associated Protein 1 (Keap1) Elisa Kit
Supplier:  Biomatik Corporation EKC34335
Encompass_Preferred
Human Kelch-Like Ech-Associated Protein 1 (Keap1) Elisa Kit for detecting Kelch-Like Ech-Associated Protein 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q14145; KEAP1. Target Synonyms: Cytosolic inhibitor of Nrf2; INrf 2; INrf2; Keap 1; KEAP1; KEAP1_HUMAN; Kelch like ECH associated protein 1; Kelch like family member 19; Kelch like protein 19; Kelch-like ECH-associated protein 1; Kelch-like protein 19; KIAA0132; KLHL 19; KLHL19; MGC10630; MGC1114; MGC20887; MGC4407; MGC9454. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4080