Learn More
Biomatik Corporation Human Kininogen-1 (Kng1) Elisa Kit
Supplier:  Biomatik Corporation EKC34353
Encompass_Preferred
Human Kininogen-1 (Kng1) Elisa Kit for detecting Kininogen 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 47-3000ng/ml. Sensitivity: 11.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01042; KNG1. Target Synonyms: Alpha-2-thiol proteinase inhibitor; BDK; BK; Bradykinin; Bradykinin included; Fitzgerald factor; FLAUJEAC FACTOR; High molecular weight kininogen; HMWK; Ile-Ser-Bradykinin; Kallidin I; Kallidin II; KNG; KNG1; KNG1_HUMAN; Low molecular weight growth-promoting factor; WILLIAMS FACTOR; Williams-Fitzgerald-Flaujeac factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4084