Learn More
Biomatik Corporation Human Laminin Subunit Alpha-4 (Lama4) Elisa Kit
Supplier:  Biomatik Corporation EKC34369
Encompass_Preferred
Human Laminin Subunit Alpha-4 (Lama4) Elisa Kit for detecting Laminin, Alpha 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.86pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q16363; LAMA4. Target Synonyms: CMD1JJ; DKFZp686D23145; LAMA3; LAMA3; formerly; LAMA4 1; LAMA4; LAMA4*-1; LAMA4_HUMAN; Laminin alpha 4; Laminin alpha 4 chain; Laminin subunit alpha 4 precursor; Laminin subunit alpha-4; Laminin-14 subunit alpha; Laminin-8 subunit alpha; Laminin-9 subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4089