Learn More
Biomatik Corporation Human Leptin Receptor, Lr/Ob-R Elisa Kit
Supplier:  Biomatik Corporation EKC34391
Encompass_Preferred
Human Leptin Receptor, Lr/Ob-R Elisa Kit for detecting Leptin Receptor in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P48357; LEPR. Target Synonyms: CD 295; CD295; CD295 antigen; Db; Fa; HuB219; LEP R; LEP-R; LEPR; LEPR_HUMAN; LEPRD; Leptin receptor; Leptin receptor fatty; Leptin receptor gene related protein; Leptin receptor precursor; Leptin receptor precursor; OB R gene related protein; OB receptor; OB-R; OB-RGRP; obl; Obr. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4093