Learn More
Biomatik Corporation Human Leukotriene E4, Lt-E4 Elisa Kit
Supplier:  Biomatik Corporation EKC34428
Encompass_Preferred
Human Leukotriene E4, Lt-E4 Elisa Kit for detecting Leukotriene E4, Lt-E4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4096