Learn More
Biomatik Corporation Human Lipoprotein Lipase (Lpl) Elisa Kit
Supplier:  Biomatik Corporation EKC34449
Encompass_Preferred
Human Lipoprotein Lipase (Lpl) Elisa Kit for detecting Lipoprotein Lipase in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06858; LPL. Target Synonyms: EC 3.1.1; EC 3.1.1.34; HDLCQ11; LIPD; LIPL_HUMAN; Lipoprotein lipase; LPL; LPL protein; MGC137861. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4100