Learn More
Biomatik Corporation Human L-Lactate Dehydrogenase B Chain (Ldhb) Elisa Kit
Supplier:  Biomatik Corporation EKC34458
Encompass_Preferred
Human L-Lactate Dehydrogenase B Chain (Ldhb) Elisa Kit for detecting Lactate Dehydrogenase B in serum, plasma, tissue homogenates, cerebrospinal fluid (csf), urine. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500mU/ml. Sensitivity: 5.8mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07195; LDHB. Target Synonyms: Epididymis secretory protein Li 281; HEL S 281; L lactate dehydrogenase B chain; L-lactate dehydrogenase B chain; Lactate Dehydrogenase B; Lactate dehydrogenase H chain; LDH B; LDH H; LDH heart subunit; LDH-B; LDH-H; LDHB; LDHB_HUMAN; LDHBD; LDHH; Renal carcinoma antigen NY REN 46; Renal carcinoma antigen NY-REN-46; TRG-5; TRG5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4102