Learn More
Biomatik Corporation Human Macrophage Colony-Stimulating Factor, M-Csf Elisa Kit
Supplier:  Biomatik Corporation EKC34514
Encompass_Preferred
Human Macrophage Colony-Stimulating Factor, M-Csf Elisa Kit for detecting Colony Stimulating Factor 1 (Macrophage) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P09603; CSF1. Target Synonyms: Colony stimulating factor 1 (macrophage); Colony stimulating factor 1; Colony stimulating factor macrophage specific; CSF 1; CSF-1; CSF1; CSF1_HUMAN; Csfm; Lanimostim; M CSF; M-CSF; Macrophage Colony Stimulating Factor 1; Macrophage colony stimulating factor; MCSF; MGC31930; Processed macrophage colony-stimulating factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4116