Learn More
Biomatik Corporation Human CAMPATH-1 antigen (CD52) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC32965
Encompass_Preferred
The Human CAMPATH-1 antigen (CD52) ELISA Kit allows for quantitative determination of CD52 molecule in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 78 ng/mL-5000 ng/mL. Sensitivity: 19.5 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P31358. Restriction: For Research Use Only.
Catalog No. 50-149-7739