Learn More
Biomatik Corporation Human Melanoma-Associated Antigen D2 (Maged2) Elisa Kit
Supplier:  Biomatik Corporation EKC34580
Encompass_Preferred
Human Melanoma-Associated Antigen D2 (Maged2) Elisa Kit for detecting Melanoma Antigen Family D, 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UNF1; MAGED2. Target Synonyms: 11B6; BCG 1; BCG-1; Breast cancer-associated gene 1 protein; HCA10; Hepatocellular carcinoma-associated protein JCL-1; JCL 1; MAGD2_HUMAN; MAGE D; MAGE D2 antigen; MAGE-D2 antigen; MAGED2; Melanoma antigen family D 2; Melanoma associated antigen D2; Melanoma-associated antigen D2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4128