Learn More
Biomatik Corporation Human Metastasis-Associated In Colon Cancer Protein 1 (Macc1) Elisa Kit
Supplier:  Biomatik Corporation EKC34596
Encompass_Preferred
Human Metastasis-Associated In Colon Cancer Protein 1 (Macc1) Elisa Kit for detecting Metastasis Associated In Colon Cancer 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 47-3000pg/ml. Sensitivity: 11.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6ZN28; MACC1. Target Synonyms: 7A5; MACC1; MACC1_HUMAN; Metastasis associated in colon cancer 1; Metastasis-associated in colon cancer protein 1; OTTHUMP00000201891; Putative binding protein 7a5; SH3 domain-containing protein 7a5; SH3BP4L. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4131