Learn More
Biomatik Corporation Human Methemoglobin,MHB ELISA Kit
Supplier:  Biomatik Corporation EKC34599
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.47 μg/mL-30 μg/mL. Sensitivity: 0.11 μg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4134