Learn More
Biomatik Corporation Human Mg7-Ag Elisa Kit
Supplier:  Biomatik Corporation EKC34606
Encompass_Preferred
Human Mg7-Ag Elisa Kit for detecting Mg7-Ag in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1-15U/ml. Sensitivity: 0.25u/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4136