Learn More
Biomatik Corporation Human Microfibrillar-Associated Protein 5 (Mfap5) Elisa Kit
Supplier:  Biomatik Corporation EKC34609
Encompass_Preferred
Human Microfibrillar-Associated Protein 5 (Mfap5) Elisa Kit for detecting Microfibrillar-Associated Protein 5(Mfap5) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800pg/ml. Sensitivity: 3.125pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13361; MFAP5. Target Synonyms: AAT9; MAGP 2; MAGP-2; MAGP2; MFAP 5; MFAP-5; MFAP5; MFAP5_HUMAN; Microfibril associated glycoprotein 2; Microfibril-associated glycoprotein 2; microfibrillar associated protein 5; Microfibrillar associated protein 5 precursor; Microfibrillar-associated protein 5; MP25. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4138