Learn More
Biomatik Corporation Human Microtubule-Associated Proteins 1A/1B Light Chain 3B (Map1Lc3B) Elisa Kit
Supplier:  Biomatik Corporation EKC34613
Encompass_Preferred
Human Microtubule-Associated Proteins 1A/1B Light Chain 3B (Map1Lc3B) Elisa Kit for detecting Microtubule-Associated Protein 1 Light Chain 3 Beta in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.67pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9GZQ8; MAP1LC3B. Target Synonyms: ATG8F; Autophagy-related protein LC3 B; Autophagy-related ubiquitin-like modifier LC3 B; LC3B; LC3II; MAP1 light chain 3 like protein 2; MAP1 light chain 3-like protein 2; MAP1A/1BLC3; MAP1A/MAP1B LC3 B; MAP1A/MAP1B light chain 3 B; MAP1ALC3; MAP1LC3B a; Map1lc3b; Microtubule associated protein 1 light chain 3 beta; Microtubule-associated protein 1 light chain 3 beta; Microtubule-associated proteins 1A/1B light chain 3B; MLP3B_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4140