Learn More
Biomatik Corporation Human Moesin (Msn) Elisa Kit
Supplier:  Biomatik Corporation EKC34636
Encompass_Preferred
Human Moesin (Msn) Elisa Kit for detecting Moesin in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P26038; MSN. Target Synonyms: Epididymis luminal protein 70; HEL70; Membrane organizing extension spike protein; Membrane-organizing extension spike protein; MOES_HUMAN; Moesin; Moesin/anaplastic lymphoma kinase fusion protein; Msn; MSN/ALK fusion. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4144