Learn More
Biomatik Corporation Human cancer antigen 27-29 (CA 27-29) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC32969
Encompass_Preferred
The Human cancer antigen 27-29 (CA 27-29) ELISA Kit allows for quantitative determination of cancer antigen 27-29 (CA 27-29) in serum. Species Reactivity: Human. Detection Range: 3 U/mL-100 U/mL. Sensitivity: 0.8 U/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7740