Learn More
Biomatik Corporation Human Ninjurin-1 (Ninj1) Elisa Kit
Supplier:  Biomatik Corporation EKC34823
Encompass_Preferred
Human Ninjurin-1 (Ninj1) Elisa Kit for detecting Ninjurin 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q92982; NINJ1. Target Synonyms: NINJ1; Ninjurin-1; Nerve injury-induced protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4171