Learn More
Biomatik Corporation Human Noggin(NOG) ELISA kit
Supplier:  Biomatik Corporation EKC34838
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q13253. Target Synonyms: SYM1, SYNS1, symphalangism 1 (proximal). Restriction: For Research Use Only.
Catalog No. 50-229-4174