Learn More
Biomatik Corporation Human Nucleophosmin (Npm1) Elisa Kit
Supplier:  Biomatik Corporation EKC34863
Encompass_Preferred
Human Nucleophosmin (Npm1) Elisa Kit for detecting Nucleophosmin (Nucleolar Phosphoprotein B23, Numatrin) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06748; NPM1. Target Synonyms: B23; MGC104254; NO38; NPM; NPM_HUMAN; NPM1; Nucleolar phosphoprotein B23; Nucleolar protein NO38; Nucleophosmin (nucleolar phosphoprotein B23 numatrin); Nucleophosmin; nucleophosmin nucleoplasmin family member 1; Nucleophosmin/nucleoplasmin family member 1; Numatrin; OTTHUMP00000161024; OTTHUMP00000161025; OTTHUMP00000223397; OTTHUMP00000223398. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4182