Learn More
Biomatik Corporation Human Nucleotide-Binding Oligomerization Domain-Containing Protein 1 (Nod1) Elisa Kit
Supplier:  Biomatik Corporation EKC34867
Encompass_Preferred
Human Nucleotide-Binding Oligomerization Domain-Containing Protein 1 (Nod1) Elisa Kit for detecting Nucleotide-Binding Oligomerization Domain Containing 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y239; NOD1. Target Synonyms: CARD 4; CARD4; Caspase recruitment domain 4; Caspase recruitment domain containing protein 4; Caspase recruitment domain family member 4; Caspase recruitment domain protein 4; Caspase recruitment domain-containing protein 4; CLR 7.1; CLR7.1; NLR family CARD domain containing 1; NLRC 1; NLRC1; NOD 1; Nod1; NOD1 protein; NOD1_HUMAN; Nucleotide binding oligomerization domain containing 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4184