Learn More
Biomatik Corporation Human Pancreatic Amylase, Pamy Elisa Kit
Supplier:  Biomatik Corporation EKC34918
Encompass_Preferred
Human Pancreatic Amylase, Pamy Elisa Kit for detecting Amylase, Alpha 2A (Pancreatic) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 47-3000mU/ml. Sensitivity: 11.75mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04746; AMY2A. Target Synonyms: AMY2A; Pancreatic alpha-amylase; PA; EC 3.2.1.1; 1; 4-alpha-D-glucan glucanohydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4198