Learn More
Biomatik Corporation Human Plasminogen Activator Inhibitor 2 (Serpinb2) Elisa Kit
Supplier:  Biomatik Corporation EKC35040
Encompass_Preferred
Human Plasminogen Activator Inhibitor 2 (Serpinb2) Elisa Kit for detecting Serpin Peptidase Inhibitor, Clade B (Ovalbumin), Member 2 in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05120; SERPINB2. Target Synonyms: HsT1201; Monocyte Arg serpin; Monocyte Arg-serpin; Monocyte Arginine-serpin; Monocyte-derived plasminogen activator inhibitor; PAI; PAI-2; PAI2; PAI2_HUMAN; Placental plasminogen activator inhibitor; PLANH2; Plasminogen activator inhibitor 2; plasminogen activator inhibitor; type 2; plasminogen activator inhibitor; type II (arginine-serpin); serine (or cysteine) proteinase inhibitor; clade B (ovalbumin); member 2; Serpin B2; serpin peptidase inhibitor; clade B (ovalbumin); member 2; Serpinb2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4217