Learn More
Biomatik Corporation Human Plexin-B1 (Plxnb1) Elisa Kit
Supplier:  Biomatik Corporation EKC35059
Encompass_Preferred
Human Plexin-B1 (Plxnb1) Elisa Kit for detecting Plexin B1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O43157; PLXNB1. Target Synonyms: PLXNB1; KIAA0407; PLXN5; SEP; Plexin-B1; Semaphorin receptor SEP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4224