Learn More
Biomatik Corporation Human Polymerase I And Transcript Release Factor (Ptrf) Elisa Kit
Supplier:  Biomatik Corporation EKC35071
Encompass_Preferred
Human Polymerase I And Transcript Release Factor (Ptrf) Elisa Kit for detecting Polymerase I And Transcript Release Factor in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 37.5-2400pg/ml. Sensitivity: 9.4pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6NZI2; CAVIN1. Target Synonyms: 2310075E07Rik; AW546441; Cavin 1; Cavin; Cavin-1; CAVIN1; CGL4; FKSG13; FLJ90031; MGC118550; OTTMUSP00000002049; Polymerase I and transcript release factor; Ptrf; PTRF_HUMAN; RNA polymerase I and transcript release factor; RP23-279L23.6; TTF I interacting peptide 12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4229