Learn More
Biomatik Corporation Human Pregnancy Zone Protein (Pzp) Elisa Kit
Supplier:  Biomatik Corporation EKC35083
Encompass_Preferred
Human Pregnancy Zone Protein (Pzp) Elisa Kit for detecting Pregnancy-Zone Protein in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 2-75ug/ml. Sensitivity: 2ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P20742; PZP. Target Synonyms: PZP; CPAMD6; Pregnancy zone protein; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4233