Learn More
Biomatik Corporation Human Pregnancy-specific beta-1-glycoprotein 9(PSG9) ELISA kit
Supplier:  Biomatik Corporation EKC35090
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 5 μg/mL-150 μg/mL. Sensitivity: 1.25 μg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q00887. Target Synonyms: PSG11, PSGII, Pregnancy-specific beta-1-glycoprotein-11|pregnancy specific beta-1-glycoprotein 11|pregnancy-specific beta-1 glycoprotein-11|pregnancy-specific beta-1-glycoprotein (PSG)|pregnancy-spe. Restriction: For Research Use Only.
Catalog No. 50-229-4235