Learn More
Biomatik Corporation Human Proadrenomedullin (Pro-Adm) Elisa Kit
Supplier:  Biomatik Corporation EKC35095
Encompass_Preferred
Human Proadrenomedullin (Pro-Adm) Elisa Kit for detecting Proadrenomedullin (Pro-Adm) in serum, plasma, cell culture supernates, urine, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20pmol/l. Sensitivity: 0.078pmol/l. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4238