Learn More
Biomatik Corporation Human Protein Dopey-2 (Dopey2) Elisa Kit
Supplier:  Biomatik Corporation EKC35194
Encompass_Preferred
Human Protein Dopey-2 (Dopey2) Elisa Kit for detecting Dopey Family Member 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 32-1200pg/ml. Sensitivity: 8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y3R5; DOPEY2. Target Synonyms: DOP1B; C21orf5; DOPEY2; KIAA0933Protein dopey-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4256