Learn More
Biomatik Corporation Human Protein Fam132A (Fam132A) Elisa Kit
Supplier:  Biomatik Corporation EKC35197
Encompass_Preferred
Human Protein Fam132A (Fam132A) Elisa Kit for detecting Family With Sequence Similarity 132, Member A in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q5T7M4; C1QTNF12. Target Synonyms: C1QTNF12; C1QDC2; CTRP12; FAM132AAdipolin; Adipose-derived insulin-sensitizing factor; C1q and TNF related protein 12; Complement C1q tumor necrosis factor-related protein 12) [Cleaved into: Adipolin fC1QTNF12; Adipolin fCTRP12; Adipolin full-length form); Adipolin gC1QTNF12; Adipolin cleaved form; Adipolin gCTRP12)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4258