Learn More
Biomatik Corporation Human Protein Kinase C-Binding Protein Nell1 (Nell1) Elisa Kit
Supplier:  Biomatik Corporation EKC35221
Encompass_Preferred
Human Protein Kinase C-Binding Protein Nell1 (Nell1) Elisa Kit for detecting Nel-Like 1 (Chicken) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q92832; NELL1. Target Synonyms: FLJ45906; IDH3GL; NEL like 1 (chicken); NEL like protein 1; Nel related protein 1; NEL-like 1 (chicken); NEL-like protein 1; Nel-related protein 1; NELL 1 protein short isoform; Nell1; NELL1_HUMAN; Neural epidermal growth factor like 1; NRP1; Protein kinase C binding protein NELL1; Protein kinase C-binding protein NELL1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4264