Learn More
Biomatik Corporation Human Protein-Glutamine Gamma-Glutamyltransferase K (Tgm1) Elisa Kit
Supplier:  Biomatik Corporation EKC35272
Encompass_Preferred
Human Protein-Glutamine Gamma-Glutamyltransferase K (Tgm1) Elisa Kit for detecting Transglutaminase 1 (K Polypeptide Epidermal Type I, Protein-Glutamine-Gamma-Glutamyltransferase) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P22735; TGM1. Target Synonyms: ARCI1; Epidermal TGase; ICR2; KTG; LI; LI1; Protein glutamine gamma glutamyltransferase K; Protein-glutamine gamma-glutamyltransferase K; TG (K); TGase 1; TGASE; TGase K; TGase-1; TGK; TGM1; TGM1_HUMAN; Transglutaminase 1 (K polypeptide epidermal type I; protein glutamine gamma glutamyltransferase); Transglutaminase 1; Transglutaminase K; Transglutaminase; keratinocyte; Transglutaminase-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4273